Stereo zoom microscopes, Stemi 305
About this item
Stemi 305 are compact Greenough stereo microscopes with 5:1 zoom for biological education, labs and industrial production environments.
- Three-dimensional and crisp images
- Integrated LED illuminations: Near vertical and oblique reflected light, as well as transmitted light
- White LEDs generate bright light of daylight colour
- Can easily be operated, also by untrained users
- Trinocular model provides a phototube for connection to any ZEISS Axiocam microscope camera
Choice between predefined microscope sets for education, industrial production and laboratory to get the optimal illumination equipment.
Stemi 305 Edu:
- LED spot, zoomable and height adjustable, for oblique and grazing light illumination with strong shadow
- Flat transmitted light base for brightfield and darkfield illumination
Stemi 305 Mat:
- LED segmentable ring light for shadow free ring illumination and oblique light segment illumination: half circle, quarter circle, two-spot
- Rotating illuminating segments
-ESD properties: anti-static coating of microscope body and stand
Stemi 305 Lab:
- Double arm gooseneck, self-carrying, for variable oblique light illumination with distinct shadow effect
- Tiltable mirror base for brightfield, darkfield and oblique light illumination
Ordering information: Eyepieces are included.
4 Options Available Below
Product Details & Documents
Stemi 305 are compact Greenough stereo microscopes with 5:1 zoom for biological education, labs and industrial production environments.
- Three-dimensional and crisp images
- Integrated LED illuminations: Near vertical and oblique reflected light, as well as transmitted light
- White LEDs generate bright light of daylight colour
- Can easily be operated, also by untrained users
- Trinocular model provides a phototube for connection to any ZEISS Axiocam microscope camera
Choice between predefined microscope sets for education, industrial production and laboratory to get the optimal illumination equipment.
Stemi 305 Edu:
- LED spot, zoomable and height adjustable, for oblique and grazing light illumination with strong shadow
- Flat transmitted light base for brightfield and darkfield illumination
Stemi 305 Mat:
- LED segmentable ring light for shadow free ring illumination and oblique light segment illumination: half circle, quarter circle, two-spot
- Rotating illuminating segments
-ESD properties: anti-static coating of microscope body and stand
Stemi 305 Lab:
- Double arm gooseneck, self-carrying, for variable oblique light illumination with distinct shadow effect
- Tiltable mirror base for brightfield, darkfield and oblique light illumination
Ordering information: Eyepieces are included.
Specifications for Product Family
Specifications
- Catalog No:
- 630-2624
- 630-2626
- 630-2625
- ZEIS4919030006000
- Observation method:
- Brightfield and darkfield
- Brightfield and darkfield
- Brightfield and darkfield
- Brightfield and darkfield
- Eyepieces:
- WF 10×/23 mm
- WF 10×/23 mm
- WF 10×/23 mm
- WF 10×/23 mm
- Zoom lens:
- 5:1
- 5:1
- 5:1
- 5:1
- Focusing system:
- Coaxial coarse and fine knobs
- Coaxial coarse and fine knobs
- Coaxial coarse and fine knobs
- Coaxial coarse and fine knobs
- Illumination:
- LED
- LED
- LED
- LED
- Camera:
- 1,2 MP
- 1,2 MP
- 1,2 MP
- 1,2 MP
Specifications
Product Family Options
Product Information
- ModelTypeMagnificationAvailabilityPrice
- 630-2624Stereo zoom microscopeSpecifications:
More
Specifications
Cat. No.630-2624Observation methodBrightfield and darkfieldEyepiecesWF 10×/23 mmZoom lens5:1Focusing systemCoaxial coarse and fine knobsIlluminationLEDCamera1,2 MP - 630-2626Stereo zoom microscopeSpecifications:
More
Specifications
Cat. No.630-2626Observation methodBrightfield and darkfieldEyepiecesWF 10×/23 mmZoom lens5:1Focusing systemCoaxial coarse and fine knobsIlluminationLEDCamera1,2 MP - 630-2625Stereo zoom microscopeSpecifications:
More
Specifications
Cat. No.630-2625Observation methodBrightfield and darkfieldEyepiecesWF 10×/23 mmZoom lens5:1Focusing systemCoaxial coarse and fine knobsIlluminationLEDCamera1,2 MP - ZEIS4919030006000Stereo zoom microscopeSpecifications:
More
Specifications
Cat. No.ZEIS4919030006000Observation methodBrightfield and darkfieldEyepiecesWF 10×/23 mmZoom lens5:1Focusing systemCoaxial coarse and fine knobsIlluminationLEDCamera1,2 MP
Accessories
Product Information
- Catalog #AvailabilityPrice
- Transport case