ContactUsLinkComponent
Aluminium seals, flip-up button top, KIMBLE®
Aluminium seals, flip-up button top, KIMBLE®
Catalog # SCERSP73843A-20
Supplier:  DWK Life Sciences
Aluminium seals, flip-up button top, KIMBLE®
Catalog # SCERSP73843A-20
Supplier:  DWK Life Sciences
Specifications
  • Type:
    Aluminium, white, unlined, flip-up button top
  • Pk:
    1.000
Specifications
About this item
Aluminium seals, unlined. Safe and easy-to-use flip off seals and allows one-handed operation.
  • With tamper evident
  • No sharp metal edges