Order Entry
Switzerland
ContactUsLinkComponent
Precast horizontal gel kits, SERVALYT™ PRECOTES™ Hp Kit
Precast horizontal gel kits, SERVALYT™ PRECOTES™ Hp Kit
Catalog # SERA42805.01
Supplier:  Serva
Precast horizontal gel kits, SERVALYT™ PRECOTES™ Hp Kit
Catalog # SERA42805.01
Supplier:  Serva

Specifications

  • Description:
    SERVALYT™ PRECOTES™ HP kit, 245×125 mm
  • Seq:
    05
  • Pk:
    1

Specifications

About this item

For haptoglobin (Hp) analysis by isoelectric focusing.

Delivery: SERVALYT™ PRECOTES™ Hp Kit contains:
5× SERVALYT™ PRECOTES™ Hp gels, 245×125 mm, 300 µm
Anode and cathode buffer solutions
Applicator strips and electrode wicks