Filter tips, OneTouch™
Supplier: Sorenson Bioscience
Certificates
About this item
Featuring a dual-shot tip design, these pipette tips combine virgin PP and pliable TPE to provide a rigid and dependable tip with a soft, easy-to-seal, color-coded collar. The innovative design creates an ergonomic tip that can be used across a wide variety of pipettors.
- Tips reduce the necessary sealing and reciprocal ejection force felt with traditional single resin PP tips
- Minimal force required to eject tip
- Ideal for multi-channel pipettors
- Certified RNase/DNase-free
- Top collar colour-coded by volume
Frequently Bought Together