Order Entry
Switzerland
ContactUsLinkComponent
pH buffers
pH buffers
Catalog # JENW16156-78
Supplier:  Cole-Parmer
CAS Number:  
undefined
pH buffers
Catalog # JENW16156-78
Supplier:  Cole-Parmer
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Description:
    pH buffer solution, 500 ml
  • Seq:
    01
  • pH value:
    9,22

Specifications

About this item

Available as single use sachets or as 500 ml bottles.