HPLC columns, SUPELCOSIL™, Suplex™ pKb-100, Supelco®
: Merck
Suplex pKb-100 columns feature the same bonded phase functionality as SUPELCOSIL LC-ABZ columns. These specially deactivated columns differ in that Suplex pKb-100 is not endcapped, while SUPELCOSIL LC-ABZ is endcapped. The absence of the end-capping reagent results in better performance from Suplex pKb-100 for the strongest basic compounds, while LC-ABZ is preferred when the sample also contains acids and/or zwitterions.
- Alkylamide phase
- Silica gel particle platform
Surface area: 170 m²/g
Carbon load: 12,5%
pH range: 2 to 7,5
Frequently Bought Together