Order Entry
Switzerland
ContactUsLinkComponent
Supelco® Ascentis® Express Phenyl-Hexyl HPLC and UHPLC Columns
Supelco® Ascentis® Express Phenyl-Hexyl HPLC and UHPLC Columns
Supplier:  Merck
Certificates
undefined
Supelco® Ascentis® Express Phenyl-Hexyl HPLC and UHPLC Columns
Supplier:  Merck

Specifications

  • Catalog No:
  • 51611-U
  • 51614-U
  • 51608-U
  • 51609-U
  • 53531-U
  • 53526-U
  • 53524-U
  • 50459-U
  • 50443-U
  • 50446-U
  • 50442-U
  • 50455-U
  • 50456-U
  • 50451-U
  • 50454-U
  • 50482-U
  • 50483-U
  • 51616-U
  • 51617-U
  • 51623-U
  • 51618-U
  • 51619-U
  • 50469-U
  • 53347-U
  • 53348-U
  • 50470-U
  • 53345-U
  • 50464-U
  • 50466-U
  • 53346-U
  • 53353-U
  • 50477-U
  • 53798-U
  • 50479-U
  • 53351-U
  • 50472-U
  • 53352-U
  • 50474-U
  • 51605-U
  • 51601-U
  • 51603-U
  • 53343-U
  • 53341-U
  • 53342-U
  • 51600-U
  • 53332-U
  • 50487-U
  • 53336-U
  • 53338-U
  • 53334-U
  • 50498-U
  • 53335-U
  • Description:
  • 2,0 µm columns
  • 2,0 µm columns
  • 2,0 µm columns
  • 2,0 µm columns
  • 2,7 µm columns
  • 2,7 µm columns
  • 2,7 µm columns
  • 5,0 µm columns
  • 5,0 µm columns
  • 5,0 µm columns
  • 5,0 µm columns
  • 5,0 µm columns
  • 5,0 µm columns
  • 5,0 µm columns
  • 5,0 µm columns
  • 5,0 µm columns
  • 5,0 µm columns
  • 2,0 µm columns
  • 2,0 µm columns
  • 2,0 µm columns
  • 2,0 µm columns
  • 2,0 µm columns
  • 5,0 µm columns
  • 2,7 µm columns
  • 2,7 µm columns
  • 5,0 µm columns
  • 2,7 µm columns
  • 5,0 µm columns
  • 5,0 µm columns
  • 2,7 µm columns
  • 2,7 µm columns
  • 5,0 µm columns
  • 2,7 µm columns
  • 5,0 µm columns
  • 2,7 µm columns
  • 5,0 µm columns
  • 2,7 µm columns
  • 5,0 µm columns
  • 2,0 µm columns
  • 2,0 µm columns
  • 2,0 µm columns
  • 2,7 µm columns
  • 2,7 µm columns
  • 2,7 µm columns
  • 2,0 µm columns
  • 2,7 µm columns
  • 5,0 µm columns
  • 2,7 µm columns
  • 2,7 µm columns
  • 2,7 µm columns
  • 5,0 µm columns
  • 2,7 µm columns
  • Application:
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Reversed-phase HPLC separations of peptides and polypeptides
  • Carbon load:
  • 6,3%
  • 6,3%
  • 6,3%
  • 6,3%
  • 7,1%
  • 7,1%
  • 7,1%
  • 5,2%
  • 5,2%
  • 5,2%
  • 5,2%
  • 5,2%
  • 5,2%
  • 5,2%
  • 5,2%
  • 5,2%
  • 5,2%
  • 6,3%
  • 6,3%
  • 6,3%
  • 6,3%
  • 6,3%
  • 5,2%
  • 7,1%
  • 7,1%
  • 5,2%
  • 7,1%
  • 5,2%
  • 5,2%
  • 7,1%
  • 7,1%
  • 5,2%
  • 7,1%
  • 5,2%
  • 7,1%
  • 5,2%
  • 7,1%
  • 5,2%
  • 6,3%
  • 6,3%
  • 6,3%
  • 7,1%
  • 7,1%
  • 7,1%
  • 6,3%
  • 7,1%
  • 5,2%
  • 7,1%
  • 7,1%
  • 7,1%
  • 5,2%
  • 7,1%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Max. pressure:
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 1000 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • pH range:
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • Surface area:
  • 120 m²/g
  • 120 m²/g
  • 120 m²/g
  • 120 m²/g
  • 135 m²/g
  • 135 m²/g
  • 135 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 120 m²/g
  • 120 m²/g
  • 120 m²/g
  • 120 m²/g
  • 120 m²/g
  • 90 m²/g
  • 135 m²/g
  • 135 m²/g
  • 90 m²/g
  • 135 m²/g
  • 90 m²/g
  • 90 m²/g
  • 135 m²/g
  • 135 m²/g
  • 90 m²/g
  • 135 m²/g
  • 90 m²/g
  • 135 m²/g
  • 90 m²/g
  • 135 m²/g
  • 90 m²/g
  • 120 m²/g
  • 120 m²/g
  • 120 m²/g
  • 135 m²/g
  • 135 m²/g
  • 135 m²/g
  • 120 m²/g
  • 135 m²/g
  • 90 m²/g
  • 135 m²/g
  • 135 m²/g
  • 135 m²/g
  • 90 m²/g
  • 135 m²/g
  • USP code:
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11

Specifications

Related Information

Guard cartridge holder

Product Information