Collect Ice (i)Xtreme Safety Ankle Boots, Lace-Up
Supplier: HONEYWELL SAFETY
Certificates
About this item
Non metallic full grain, leather ankle boot with a strong water resistant upper construction and an innovative PU/nitrile outsole for extrem performances.
- (i)NTELLIGENT COLD INSULATION: For a strong cold insulation
- (i)NTEGRATED ANKLE PROTECTION: For additional ankle protection
- (i)NTELLIGENT WEAR CONTROL: Helps users to detect when the outsole is used
- Bi-density insole for improved comfort
- Non metallic anti-puncture midsole
- Smartium non metallic toecap made from injected polymer
Colour: Brown
EN ISO 20345:2011; S3, SRC, HI, HRO, CI, AN.
Frequently Bought Together