ContactUsLinkComponent
Gas purifier, 73-099 AA
Gas purifier, 73-099 AA
Catalog # PERCN9301398
Supplier:  PerkinElmer
Gas purifier, 73-099 AA
Catalog # PERCN9301398
Supplier:  PerkinElmer
Specifications
  • Description:
    Gas purifier, 73-099 AA
  • Gases purified:
    Compressed air and acetylene gases
  • Pk:
    1
Specifications
About this item
This wall mounted gas purifier system is designed to purify compressed air and acetylene gas.
It consists of two independent filtration systems, one for compressed air and one for acetylene. The unit also has a flashback arrestor on the acetylene line and a pressure regulator on the compressed air line.
The two-stage air filtration assembly consists of a Balston grade DX coalescing filter and a Balston grade BX coalescing filter. These filters remove oil, water and particulate contamination (99,99% at 0,01 micron) from the compressed air supply.