ContactUsLinkComponent
Reusable earplugs, E-A-R™ Ultrafit™ X
Reusable earplugs, E-A-R™ Ultrafit™ X
Catalog # 111-1629
Supplier:  3M
undefined
Reusable earplugs, E-A-R™ Ultrafit™ X
Catalog # 111-1629
Supplier:  3M
Specifications
  • Colour:
    Yellow/blue
  • Description:
    Earplugs, with cord, 35 dB, with storage box
  • Seq:
    02
  • Size:
    U*
  • Pk:
    50 PAIR
Specifications
About this item
Corded earplugs made from a patented polymer formulation, the comfortable triple flanged tip of these earplugs provides an excellent attenuation of 35 dB SNR, one of the highest amongst pre-moulded earplugs. The earplugs are already pre-formed to ensure optimum fit.
  • Triple-flange feature of the earplugs ensures a perfect fit: No size adjustment required
  • Fit most ear canals comfortably and provide a high level of attenuation
  • Moisture resistant and compatible with other PPE
  • Washable and reusable
SNR: 35 dB
Certifications: EN 352-2