Horizontal laminar flow, Purifier®
Supplier: LABCONCO
Certificates
About this item
It offers a choice to protect materials that do not generate harmful aerosols or vapors. Its been designed with 99,99% efficient HEPA filter, the air from this minimizes cross contamination in the work area and provides a particulate free work environment. It has wide range of applications including plant tissue culture research, media preparation, electronic part inspection, syringe filling, medical device assembly and non hazardous parenteral drug formulation. These benches should never be used in conjunction with biohazardous material, toxins or radionuclides.
- Differential pressure gauge
- Fluorescent lighting
- Glacier white powder coated steel exterior
- Available in various width sizes and has a differential pressure gauge that displays pressure across the HEPA filter
Construction: Work surface made of stainless steel
Certifications: Air quality Class 5 - ISO 14644-1 (Class 100).
Specifications
- Catalog No:
- LBCP3873029
- LBCP3889708
- LBCP3889709
- LBCP3873028
- LBCP3873009
- LBCP3888428
- LBCP3888429
- LBCP3889729
- LBCP3888408
- LBCP3873008
- LBCP3889728
- Model:
- Purifier®
- Purifier®
- Purifier®
- Purifier®
- Purifier®
- Purifier®
- Purifier®
- Purifier®
- Purifier®
- Purifier®
- Purifier®
- Lighting:
- Fluorescent lighting
- Fluorescent lighting
- Fluorescent lighting
- Fluorescent lighting
- Fluorescent lighting
- Fluorescent lighting
- Fluorescent lighting
- Fluorescent lighting
- Fluorescent lighting
- Fluorescent lighting
- Fluorescent lighting
- Filters:
- HEPA filter: 99,99% efficient
- HEPA filter: 99,99% efficient
- HEPA filter: 99,99% efficient
- HEPA filter: 99,99% efficient
- HEPA filter: 99,99% efficient
- HEPA filter: 99,99% efficient
- HEPA filter: 99,99% efficient
- HEPA filter: 99,99% efficient
- HEPA filter: 99,99% efficient
- HEPA filter: 99,99% efficient
- HEPA filter: 99,99% efficient
- Electrical supply:
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz
- Ext. W×D×H:
- 1905×991×1229 mm
- 965×864×1229 mm
- 965×991×1229 mm
- 1905×864×1229 mm
- 1880×991×1229 mm
- 1295×991×1229 mm
- 1295×864×1229 mm
- 991×991×1229 mm
- 1270×864×1229 mm
- 1880×864×1229 mm
- 991×864×1229 mm
- Weight:
- 299 kg
- 165 kg
- 169 kg
- 293 kg
- 299 kg
- 206 kg
- 211 kg
- 169 kg
- 206 kg
- 293 kg
- 165 kg