Order Entry
Switzerland
ContactUsLinkComponent
Anti-Thryoid Stimulating Hormone (TSH, intact) Goat Polyclonal Antibody
Anti-Thryoid Stimulating Hormone (TSH, intact) Goat Polyclonal Antibody
Catalog # IMMRIR2358
Supplier:  ImmunoReagents
Anti-Thryoid Stimulating Hormone (TSH, intact) Goat Polyclonal Antibody
Catalog # IMMRIR2358
Supplier:  ImmunoReagents

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Thyroid Stimulating Hormone
  • Antigen symbol:
    TSH
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Purified IgG
  • Form:
    Clear, colorless liquid, 0,2 µm filtered
  • Antigen synonyms:
    follicle-stimulating hormone alpha chain|follicle-stimulating hormone alpha subunit|chorionic gonadotrophin subunit alpha|choriogonadotropin alpha chain|follitropin alpha chain|luteinizing hormone alpha chain|glycoprotein hormones alpha chain|lutropin alpha chain|FSH-alpha|thyrotropin alpha chain|thyroid-stimulating hormone alpha chain|LSH-alpha|alpha polypeptide|TSH-alpha|anterior pituitary glycoprotein hormones common subunit alpha|chorionic gonadotropin
  • Storage buffer:
    10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2
  • Storage temperature:
    2...8 °C
  • Concentration:
    > 4,5 mg/ml (E 1% at 280 nm = 13,0)
  • Shipping temperature:
    2...8 °C
  • Immunogen:
    Human Thryoid Stimulating Hormone (TSH, intact)
  • Tested applications:
    Fluorescence-Activated Cell Sorting, Immunodiffusion, Labeling
  • Purification:
    Affinity purified using solid phase Human Thyroid Stimulating Hormone (TSH, intact)
  • Size:
    0,5 mg
  • Pk:
    1 mg

Specifications

About this item

ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy.  Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.  

  • Origin: US

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

ImmunoReagents antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002.
Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Primary
Antigen: Thryoid Stimulating Hormone (TSH, intact)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human