Anti-Sulfaquinoxaline Sheep Polyclonal Antibody
Catalog # USBIS8030-06
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Sulfaquinoxaline
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- Form:liquid
- Storage buffer:20mM PBS, 0,09% sodium azide, pH 7,2.
- Storage temperature:Cool
- Concentration:~~1 mg/ml (UV abs)
- Shipping temperature:Ice
- Purification:purified
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: ELISA: 10 ?g/ml
Type: Primary
Antigen: Sulfaquinoxaline
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: