Anti-CCL5 Rabbit polyclonal antibody
USBIR1100-14-100
- Antibody type:Primary
- Antigen name:chemokine (C-C motif) ligand 5
- Antigen symbol:CCL5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Neutralisation:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: CCL5 (chemokine (C-C motif) ligand 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse