Order Entry
Switzerland
ContactUsLinkComponent
Anti-Protein Gene Product 9.5 Mouse Monoclonal Antibody [clone: 9i250]
Anti-Protein Gene Product 9.5 Mouse Monoclonal Antibody [clone: 9i250]
Catalog # USBIP9102-71E
Supplier:  US Biological
Anti-Protein Gene Product 9.5 Mouse Monoclonal Antibody [clone: 9i250]
Catalog # USBIP9102-71E
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein Gene Product 9.5
  • Clonality:
    Monoclonal
  • Clone:
    9i250
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Bovine,
    Rat
  • Western blot:
    Yes
  • Format:
    Supernatant
  • Form:
    liquid
  • Storage buffer:
    10mMsodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Size:
    500 µl
  • Pk:
    500 µl

Specifications

About this item

Recommended dilution: Immunofluorescence: 1:1000; Western Blot: 1:10000

Type: Primary
Antigen: Protein Gene Product 9.5
Clonality: monoclonal
Clone: 9i250
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Rat