Anti-Protein Gene Product 9.5 Rabbit Polyclonal Antibody
USBIP9102-70F
- Antibody type:Primary
- Antigen name:Protein Gene Product 9.5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Hamster,Pig,Rabbit,Sheep,Human,Bovine,Guinea pig,Dog,Rat,Mouse,Cat
- Western blot:Yes
- Format:Serum
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
- Recommended dilution: Immunochemistry: 1:200-1:800
Type: Primary
Antigen: Protein Gene Product 9.5
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Cat, Dog, Guinea pig, Hamster, Human, Mouse, Pig, Rabbit, Rat, Sheep