Anti-Progesterone Mouse Monoclonal Antibody [clone: 10F313]
Catalog # USBIP9006-01A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Progesterone
- Clonality:Monoclonal
- Clone:10F313
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Recommended dilution: ELISA: 1:5000-1:15000
Type: Primary
Antigen: Progesterone
Clonality: monoclonal
Clone: 10F313
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: