Anti-SLC5A5 Mouse Monoclonal Antibody [clone: 12F4]
Catalog # USBIN2750-10B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 5 (sodium/iodide cotransporter), member 5
- Antigen symbol:Solute carrier family 5, member 5
- Clonality:Monoclonal
- Clone:12F4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:468 to 643
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: SLC5A5 (solute carrier family 5 (sodium/iodide cotransporter), member 5)
Clonality: monoclonal
Clone: 12F4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat