Anti-MEF2C Rabbit Polyclonal Antibody
Catalog # USBIM9769-97C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Myocyte enhancer factor 2C
- Antigen symbol:MEF2C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Modification:phospho-Ser396
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Dot Blot
- Purification:purified by immunoaffinity chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000 Dot Blot: 1:100-1:500
Type: Primary
Antigen: MEF2C (myocyte enhancer factor 2C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human