Anti-MHC Class 1 Chain-related Gene A, B Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10F158]
Catalog # USBIM3886-75Y3
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:MHC Class 1 Chain-related Gene A, B
- Clonality:Monoclonal
- Clone:10F158
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,4, 1% BSA, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: MHC Class 1 Chain-related Gene A, B
Clonality: monoclonal
Clone: 10F158
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human