Order Entry
Switzerland
ContactUsLinkComponent
Anti-MHC Class 1 Chain-related Gene A Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anti-MHC Class 1 Chain-related Gene A Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Catalog # USBIM3886-75A-APC
Supplier:  US Biological
undefined
Anti-MHC Class 1 Chain-related Gene A Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Catalog # USBIM3886-75A-APC
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MHC Class 1 Chain-related Gene A
  • Clonality:
    Polyclonal
  • Conjugation:
    APC (Allophycocyanin)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    75 to 105
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    purified by Protein A chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100; Flow Cytometry: 1:10-1:50; Immunofluorescence: 1:10-1:50
Do not freeze!

Type: Primary
Antigen: MHC Class 1 Chain-related Gene A
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human