Anti-Hepatitis C Virus Core antigen Goat Polyclonal Antibody
USBIH1920-08E
- Antibody type:Primary
- Antigen name:Hepatitis C Virus Core antigen
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~4-5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:1 ml
- Pk:1 mL
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: Hepatitis C Virus Core antigen
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: