Anti-GRIA2 Rabbit Polyclonal Antibody
Catalog # USBIG3500-18
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Glutamate receptor 2
- Antigen symbol:GRIA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:873 to 883
- Modification:phospho-Ser880
- Storage buffer:0,2M Tris-glycine, pH 7,4, 0,15M sodium chloride, 0,05% sodium azide, before the addition of glycerol to 30%.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000-1:10000
Type: Primary
Antigen: GRIA2 (glutamate receptor, ionotropic, AMPA 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat