Order Entry
Switzerland
ContactUsLinkComponent
Anti-Diethylstilbestrol Sheep polyclonal antibody
  USBID3551-34B
Anti-Diethylstilbestrol Sheep polyclonal antibody
  USBID3551-34B

 

  • Antibody type:
    Primary
  • Antigen name:
    Diethylstilbestrol
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Sheep
  • Form:
    liquid
  • Storage buffer:
    20mM PBS, pH 7,2, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Recommended dilution: ELISA: 0.156?g/ml

Type: Primary
Antigen: Diethylstilbestrol
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: