Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-Corticotropin-releasing Factor Receptor 2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
  USBIC7903-90D4
Anti-Corticotropin-releasing Factor Receptor 2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
  USBIC7903-90D4

 

  • Antibody type:
    Primary
  • Antigen name:
    Corticotropin releasing factor receptor 2
  • Antigen symbol:
    CRF2
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    liquid
  • Amino acid number:
    385 to 411
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    purified
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: Corticotropin-releasing Factor Receptor 2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human