Auftragserfassung
Switzerland
ContactUsLinkComponent
Anti-GJA5 Mouse Monoclonal Antibody [clone: 6A47]
Katalognr. USBIC7856-03B
Anbieter:  US Biological
Anti-GJA5 Mouse Monoclonal Antibody [clone: 6A47]
Katalognr. USBIC7856-03B
Anbieter:  US Biological

Technische Daten

  • Antibody type:
    Primary
  • Antigen name:
    gap junction Protein, alpha 5, 40kDa
  • Antigen symbol:
    GJA5
  • Clonality:
    Monoclonal
  • Clone:
    6A47
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-terminal
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Technische Daten

Über diesen Artikel

Recommended dilution: Western Blot: 1-3?g/ml; Immunochemistry: 1-3?g/ml; ELISA: 0.1-1 ?g/ml

Type: Primary
Antigen: GJA5 (gap junction protein, alpha 5, 40kDa)
Clonality: Monoclonal
Clone: 6A47
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse