Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-C9 Mouse monoclonal antibody [clone: 64E9]
  USBIC7850-53D
undefined
Anti-C9 Mouse monoclonal antibody [clone: 64E9]
  USBIC7850-53D

 

  • Antibody type:
    Primary
  • Antigen name:
    complement component 9
  • Antigen symbol:
    C9
  • Clonality:
    Monoclonal
  • Clone:
    64E9
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    HEPES, 0,15M sodium chloride, 0,01% BSA, 0,03% sodium azide, 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by ammonium sulfate precipitation
  • Size:
    50 µl
  • Pk:
    50 µl

 

 

Recommended dilution: Western Blot: 1:2000; Immunochemistry: 1:200

Type: Primary
Antigen: C9 (complement component 9)
Clonality: Monoclonal
Clone: 64000000000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human