Anti-COL4 Mouse Monoclonal Antibody [clone: 5F75]
Catalog # USBIC7510-50A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:collagen, type IV
- Antigen symbol:COL4
- Clonality:Monoclonal
- Clone:5F75
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Format:ascites
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Primary
Antigen: COL4 (collagen, type IV)
Clonality: Monoclonal
Clone: 5F75
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human