Order Entry
Switzerland
ContactUsLinkComponent
Anti-COL3 Rabbit Polyclonal Antibody
Anti-COL3 Rabbit Polyclonal Antibody
Catalog # USBIC7510-39X
Supplier:  US Biological
undefined
Anti-COL3 Rabbit Polyclonal Antibody
Catalog # USBIC7510-39X
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    collagen, type III
  • Antigen symbol:
    COL3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rabbit,
    Mammals,
    Human,
    Bovine,
    Rat
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    0,125M sodium borate, 0,075M sodium chloride, 0,005M EDTA, 0,1% sodium azide, pH 8.0
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: ELISA: 1:5000-1:50000 Immunochemistry: 1:20-1:200; Immunoprecipitation: 1:100; Western Blot: 1:1000-1:10000

Type: Primary
Antigen: COL3 (collagen, type III)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Mammalian, Rabbit, Rat