Anti-COL2A1 Rabbit Polyclonal Antibody (Biotin)
USBIC7510-37G-BIOT
- Antibody type:Primary
- Antigen name:collagen, type II, alpha 1
- Antigen symbol:COL2A1
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: COL2A1 (collagen, type II, alpha 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human