Anti-ITGA6 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H164]
USBIC2405-04R
- Antibody type:Primary
- Antigen name:Integrin Alpha 6
- Antigen symbol:ITGA6
- Clonality:Monoclonal
- Clone:7H164
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 15% glycerol, 0,2% BSA, 0,05% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:120 Tests
- Pk:120 Tests
Do not freeze!
Type: Primary
Antigen: ITGA6 (integrin, alpha 6)
Clonality: Monoclonal
Clone: 7H164
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human