Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-Calcitonin Gene-Related Peptide Rabbit Polyclonal Antibody
  USBIC0115-02V
Anti-Calcitonin Gene-Related Peptide Rabbit Polyclonal Antibody
  USBIC0115-02V

 

  • Antibody type:
    Primary
  • Antigen name:
    Calcitonin Gene Related Peptide
  • Antigen symbol:
    CGRP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Rat
  • Format:
    Serum
  • Form:
    lyophilized powder
  • Storage buffer:
    0,1ml of 10 mg/ml BSA is PBS
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: Calcitonin Gene-Related Peptide
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat