Order Entry
Switzerland
ContactUsLinkComponent
Anti-Calcitonin Gene-Related Peptide Chicken Polyclonal Antibody
Anti-Calcitonin Gene-Related Peptide Chicken Polyclonal Antibody
  USBIC0115-01G
Anti-Calcitonin Gene-Related Peptide Chicken Polyclonal Antibody
  USBIC0115-01G

 

  • Antibody type:
    Primary
  • Antigen name:
    Calcitonin Gene Related Peptide
  • Antigen symbol:
    CGRP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Chicken
  • Isotype:
    IgY
  • Reactivity:
    Rat
  • Format:
    Serum
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pk:
    50 µl

 

 

Recommended dilution: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: Calcitonin Gene-Related Peptide
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Rat