Anti-Calcitonin Gene Related Peptide Goat polyclonal antibody unconjugated
USBIC0115-01D
- Antibody type:Primary
- Antigen name:Calcitonin Gene Related Peptide
- Antigen symbol:CGRP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Form:liquid
- Amino acid number:23 to 37
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µl
- Pk:100 µl
Recommended dilution: ELISA: 1:500-1:2500
Type: Primary
Antigen: Calcitonin Gene Related Peptide
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat