Anti-BCAS3 Rabbit polyclonal antibody unconjugated
Catalog # USBIB0395-60D
Anbieter: US Biological
Technische Daten
- Antibody type:Primary
- Antigen name:breast carcinoma amplified sequence 3
- Antigen symbol:BCAS3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Technische Daten
Über diesen Artikel
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: BCAS3 (breast carcinoma amplified sequence 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human