Order Entry
Switzerland
ContactUsLinkComponent
Anti-AMP Activated Protein Kinase 1, 2 Rabbit Polyclonal Antibody
Anti-AMP Activated Protein Kinase 1, 2 Rabbit Polyclonal Antibody
Catalog # USBIA1475-08E
Supplier:  US Biological
Anti-AMP Activated Protein Kinase 1, 2 Rabbit Polyclonal Antibody
Catalog # USBIA1475-08E
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    AMP Activated Protein Kinase 1/2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Modification:
    phospho-Ser485, phospho-Ser491
  • Storage buffer:
    PBS, pH 7,4, 150mM sodium chloride, 0,02% sodium azide and 50% glycerol.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: AMP Activated Protein Kinase 1, 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human