Anti-PRKAB2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # USBIA1475-03J1FITC
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Protein Kinase AMP-Activated Non-Catalytic Subunit Beta 2
- Antigen symbol:PRKAB2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: PRKAB2 (protein kinase, AMP-activated, beta 2 non-catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse