Anti-Protein Gene Product 9.5 Mouse Monoclonal Antibody [clone: 31A3]
Katalognr. USBI138591-100
Anbieter: US Biological
Technische Daten
- Antibody type:Primary
- Antigen name:Protein Gene Product 9.5
- Clonality:Monoclonal
- Clone:31A3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Pig,Human,Bovine,Rat,Mouse
- Form:liquid
- Storage buffer:PBS, 0,05% BSA, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,2 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Technische Daten
Über diesen Artikel
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 0.5-1 ?g/ml
Type: Primary
Antigen: Protein Gene Product 9.5
Clonality: monoclonal
Clone: 31A3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Bovine, Human, Mouse, Pig, Rat