Anti-CD5 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 13H170]
N° de catalogue USBI137761
Fournisseur: US Biological
Spécifications
- Antibody type:Primary
- Antigen name:CD5 molecule
- Antigen symbol:CD5
- Clonality:Monoclonal
- Clone:13H170
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS,sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Spécifications
À propos de cet article
Do not freeze!
Type: Primary
Antigen: CD5 (CD5 molecule)
Clonality: monoclonal
Clone: 13H170
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse