Anti-Plasmodium falciparum, HRP2 Mouse Monoclonal Antibody [clone: 13F172]
Catalog # USBI136714
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Plasmodium falciparum
- Antigen symbol:HRP2
- Clonality:Monoclonal
- Clone:13F172
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human,Mouse
- Form:liquid
- Storage buffer:0,1MPBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Plasmodium falciparum, HRP2
Clonality: monoclonal
Clone: 13F172
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse