Anti-RNF13 Mouse Monoclonal Antibody [clone: 3E4]
Catalog # USBI132646
Anbieter: US Biological
Technische Daten
- Antibody type:Primary
- Antigen name:Ring Finger Protein 13
- Antigen symbol:RNF13
- Clonality:Monoclonal
- Clone:3E4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:51 to 151
- Storage buffer:PBS, pH 7,2.
- Sequence:LPARFGYRLPAEGLKGFLINSKPENACEPIVPPPVKDNSSGTFIVLIRRLDCNFDIKVLNAQRAGYKAAIVHNVDSDDLISMGSNDIEVLKKIDIPSVFI*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: RNF13 (ring finger protein 13)
Clonality: monoclonal
Clone: 30000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human