Anti-PRKAG2 Mouse Monoclonal Antibody [clone: 3C4]
Katalognr. USBI131792
Anbieter: US Biological
Technische Daten
- Antibody type:Primary
- Antigen name:Protein Kinase AMP-Activated Non-Catalytic Subunit Gamma 2
- Antigen symbol:PRKAG2
- Clonality:Monoclonal
- Clone:3C4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:AFMKQNLDELGIGTYHNIAFIHPDTPIIKALNIFVERRISALPVVDESGKVVDIYSKFDVINLAAEKTYNNLDITVTQALQHRSQYFEGVVKCNKLEILETIVDRIVRAE
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Technische Daten
Über diesen Artikel
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: PRKAG2 (protein kinase, AMP-activated, gamma 2 non-catalytic subunit)
Clonality: monoclonal
Clone: 3C4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human