Anti-PAGE4 Mouse Monoclonal Antibody [clone: 7C3]
Catalog # USBI130832
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:P antigen family, member 4 (prostate associated)
- Antigen symbol:PAGE4
- Clonality:Monoclonal
- Clone:7C3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 102
- Storage buffer:PBS, pH 7,2.
- Sequence:MSARVRSRSRGRGDGQEAPDVVAFVAPGESQQEEPPTDNQDIEPGQEREGTPPIEERKVEGDCQEMDLEKTRSERGDGSDVKEKTPPNPKHAKTKEAGDGQP
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: PAGE4 (P antigen family, member 4 (prostate associated))
Clonality: monoclonal
Clone: 7C3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse