Anti-GRIA1 Mouse Monoclonal Antibody [clone: 3D10]
USBI127363
- Antibody type:Primary
- Antigen name:Glutamate receptor 1
- Antigen symbol:GRIA1
- Clonality:Monoclonal
- Clone:3D10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:liquid
- Amino acid number:151 to 250
- Storage buffer:PBS, pH 7,2.
- Sequence:SLLGHYKWEKFVYLYDTERGFSILQAIMEAAVQNNWQVTARSVGNIKDVQEFRRIIEEMDRRQEKRYLIDCEVERINTILEQVVILGKHSRGYHYMLANL
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: GRIA1 (glutamate receptor, ionotropic, AMPA 1)
Clonality: monoclonal
Clone: 3D10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human