Anti-GDF3 Mouse Monoclonal Antibody [clone: 2B10]
Catalog # USBI127253
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Growth differentiation factor 3
- Antigen symbol:GDF3
- Clonality:Monoclonal
- Clone:2B10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Form:liquid
- Amino acid number:265 to 364
- Storage buffer:PBS, pH 7,2.
- Sequence:HRHQLFINFRDLGWHKWIIAPKGFMANYCHGECPFSLTISLNSSNYAFMQALMHAVDPEIPQAVCIPTKLSPISMLYQDNNDNVILRHYEDMVVDECGCG
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: GDF3 (growth differentiation factor 3)
Clonality: monoclonal
Clone: 2B10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human