Anti-ANGPTL4 Mouse Monoclonal Antibody [clone: Kairos4-153AD]
Katalognr. USBI044530
Anbieter: US Biological
Technische Daten
- Antibody type:Primary
- Antigen name:Angiopoietin-like 4
- Antigen symbol:Aminomethyltransferase
- Clonality:Monoclonal
- Clone:Kairos4-153AD
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Western blot:Yes
- Form:liquid
- Antigen synonyms:ANGPTL4
- Storage buffer:0,2um-filtered PBS, pH 7,4
- Storage temperature:Cool
- Concentration:0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: ANGPTL4 (angiopoietin-like 4)
Clonality: Monoclonal
Clone: Kairos4-153AD
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: