Order Entry
Switzerland
ContactUsLinkComponent
Anti-PLCL2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anti-PLCL2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Catalog # USBI040142-APC
Supplier:  US Biological
undefined
Anti-PLCL2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Catalog # USBI040142-APC
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Phospholipase C-like 2
  • Antigen symbol:
    PLCL2
  • Clonality:
    Polyclonal
  • Conjugation:
    APC (Allophycocyanin)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Form:
    liquid
  • Amino acid number:
    1100 to 1127
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    purified by Protein A chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50; Immunofluorescence: 1:10-50; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: PLCL2 (phospholipase C-like 2)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human