Order Entry
Switzerland
ContactUsLinkComponent
Anti-NEDD4L Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-NEDD4L Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # USBI038877-FITC
Anbieter:  US Biological
undefined
Anti-NEDD4L Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # USBI038877-FITC
Anbieter:  US Biological

Technische Daten

  • Antibody type:
    Primary
  • Antigen name:
    Neural Precursor Cell Expressed, Developmentally Down-Regulated 4-Like, E3 Ubiquitin Protein Ligase
  • Antigen symbol:
    NEDD4L
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    internal
  • Form:
    liquid
  • Amino acid number:
    575 to 604
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    purified by Protein A chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

Technische Daten

Über diesen Artikel

Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: NEDD4L (neural precursor cell expressed, developmentally down-regulated 4-like, E3 ubiquitin protein ligase)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse