Anti-p21 WAF1 (Ab-1) (EA10) Mouse Monoclonal Antibody [clone: EA10]
Katalognr. OP64-100
Anbieter: Merck Millipore (Oncogene)
Technische Daten
- Antibody type:Primary
- Antigen symbol:P21
- Clonality:Monoclonal
- Clone:EA10
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:WAF1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: p21 WAF1 (Ab-1) (EA10)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human