Anti-c-Fos (Ab-1) (2G9C3) Mouse Monoclonal Antibody [clone: 2G9C3]
Catalog # OP17-200
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen symbol:cFOS
- Clonality:Monoclonal
- Clone:2G9C3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: c-Fos (Ab-1) (2G9C3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: